accessibility menu, dialog, popup

UserName

CAS RN 620112-78-9

CH3OONNH3CNNS(E/Z)

CAS RN 620112-78-9

IUPAC Name: 2-(1,3-benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl]prop-2-enenitrile
Synonyms: 2-(1,3-benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxopyrido[1,2-a]pyrimidin-3-yl]prop-2-enenitrile
Molecular Weight (g/mol): 450.52
Molecular Formula: C26H18N4O2S
InChi Key: VFSVKVQMZDJFQX-UHFFFAOYSA-N
SMILES: CC1=CC(OC2=C(C=C(C#N)C3=NC4=CC=CC=C4S3)C(=O)N3C=CC=C(C)C3=N2)=CC=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results