accessibility menu, dialog, popup

UserName

HOOHO

CAS RN 621-54-5

IUPAC Name: 3-(3-hydroxyphenyl)propanoic acid
Synonyms: 3-hydroxyphenylpropionic acid dihydro-m-coumaric acid dihydro-3-coumaric acid 3-hydroxydihydrocinnamate dihydro-m-coumarate 3-hydroxydihydrocinnamic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: QVWAEZJXDYOKEH-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)