CAS RN 6212-33-5
CAS RN 6212-33-5
IUPAC Name:
(2R)-2-azaniumyl-2-(4-chlorophenyl)acetate
Synonyms:
(2R)-2-ammonio-2-(4-chlorophenyl)acetate
Molecular Weight (g/mol):
185.61
Molecular Formula:
C8H8ClNO2
InChi Key:
QGJGBYXRJVIYGA-SSDOTTSWSA-N
SMILES:
[NH3+][C@@H](C([O-])=O)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
4-Chloro-DL-phenylglycine, 98%
CAS: 6212-33-5 Molecular Formula: C8H8ClNO2 Molecular Weight (g/mol): 185.607 MDL Number: MFCD00049328 InChI Key: QGJGBYXRJVIYGA-UHFFFAOYSA-N Synonym: 2-amino-2-4-chlorophenyl acetic acid,amino 4-chlorophenyl acetic acid,amino-4-chloro-phenyl-acetic acid,dl-2-4-chlorophenyl glycine,2-4-chlorophenyl glycine,+/--4-chlorophenylglycine,h-dl-phg 4-cl-oh,4-chloro-dl-phenylglycine,dl-4-chlorophenyl glycine,4-chlorophenyl glycine PubChem CID: 587821 IUPAC Name: 2-amino-2-(4-chlorophenyl)acetic acid SMILES: C1=CC(=CC=C1C(C(=O)O)N)Cl