missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-DL-phenylglycine, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 0.1mol/L NaOH (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 0.1mol/L NaOH (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Color | White |
| Solubility Information | Soluble in 0.1mol/L NaOH (almost transparency). |
| Quantity | 100 g |
| Formula Weight | 185.61 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 4-Chloro-DL-phenylglycine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction