accessibility menu, dialog, popup

UserName

CH3NHNH2O

CAS RN 622-51-5

IUPAC Name: (4-methylphenyl)urea
Synonyms: 4-tolylurea
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: DMSHKWHLXNDUST-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(N)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results