accessibility menu, dialog, popup

UserName

BrN

CAS RN 623-00-7

IUPAC Name: 4-bromobenzonitrile
Synonyms: 4-bromobenzonitrile
Molecular Weight (g/mol): 182.02
Molecular Formula: C7H4BrN
InChi Key: HQSCPPCMBMFJJN-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C#N
Quantity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)