accessibility menu, dialog, popup

UserName

CAS RN 623-48-3

IUPAC Name: ethyl 2-iodoacetate
Synonyms: ethyl iodoacetate
Molecular Weight (g/mol): 214
Molecular Formula: C4H7IO2
InChi Key: MFFXVVHUKRKXCI-UHFFFAOYSA-N
SMILES: CCOC(=O)CI
Boiling Point 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)