Product Code. 10656594
Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10656594
250 mL
Glass bottle
250mL
10501861
10 mL
Glass bottle
10mL
10338120
50 mL
Glass bottle
50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10656594
Supplier
Thermo Scientific
Supplier No.
222192500
Estimated Shipment: 17-11-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
623-48-3
Molecular Formula
C4H7IO2
Molecular Weight (g/mol)
214.00
MDL Number
MFCD00001081
InChI Key
MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym
ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID
12183
IUPAC Name
ethyl 2-iodoacetate
SMILES
CCOC(=O)CI
Specifications
Color
Orange to Yellow
Density
1.8000g/mL
Boiling Point
179°C to 180°C
Flash Point
76°C
Infrared Spectrum
Authentic
Assay Percent Range
97.5% min. (GC)
Packaging
Glass bottle
Linear Formula
ICH2 CO2 C2 H5
Refractive Index
1.5035 to 1.5055
Quantity
250 mL
Beilstein
02,222
Specific Gravity
1.8
Solubility Information
Solubility in water: immiscible
Stabilizer
copper
Formula Weight
214
Percent Purity
98%
Physical Form
Liquid
Chemical Name or Material
Ethyl iodoacetate, 98%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?