accessibility menu, dialog, popup

UserName

CAS RN 6232-11-7

CH3OONH2HCl

CAS RN 6232-11-7

IUPAC Name: methyl 4-(aminomethyl)benzoate hydrochloride
Synonyms: methyl 4-(aminomethyl)benzoate hydrochloride
Molecular Weight (g/mol): 201.65
Molecular Formula: C9H12ClNO2
InChi Key: GIZCKBSSWNIUMZ-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1=CC=C(CN)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results