accessibility menu, dialog, popup

UserName

CH3SH3C

CAS RN 624-89-5

IUPAC Name: (methylsulfanyl)ethane
Synonyms: ethyl methyl sulfide
Molecular Weight (g/mol): 76.16
Molecular Formula: C3H8S
InChi Key: WXEHBUMAEPOYKP-UHFFFAOYSA-N
SMILES: CCSC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results