accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 62451-84-7

IUPAC Name: methyl 2-[3-(trifluoromethyl)phenyl]acetate
Synonyms: methyl 2-[3-(trifluoromethyl)phenyl]acetate
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: GMEBDKGPPKAPEM-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results