accessibility menu, dialog, popup

UserName

NH2ClFFF

CAS RN 62476-58-8

IUPAC Name: 2-chloro-3-(trifluoromethyl)aniline
Synonyms: 2-chloro-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 195.57
Molecular Formula: C7H5ClF3N
InChi Key: WLXOMMTWPMQFDE-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1Cl)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results