accessibility menu, dialog, popup

UserName

CAS RN 625-35-4

CH3ClO(E)

CAS RN 625-35-4

IUPAC Name: (2E)-but-2-enoyl chloride
Synonyms: 2-butenoyl chloride
Molecular Weight (g/mol): 104.53
Molecular Formula: C4H5ClO
InChi Key: RJUIDDKTATZJFE-NSCUHMNNSA-N
SMILES: C\C=C\C(Cl)=O
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)