missing translation for 'onlineSavingsMsg'
Learn More

trans-Crotonyl chloride, 90%, tech.

Product Code. 10321344 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10321344

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-35-4 | C4H5ClO | 104.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-35-4
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.54
MDL Number MFCD00000734
InChI Key RJUIDDKTATZJFE-NSCUHMNNSA-N
Synonym crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride
PubChem CID 643131
IUPAC Name (E)-but-2-enoyl chloride
SMILES CC=CC(=O)Cl

Specifications

Color Colorless to Brown
Density 1.0900g/mL
Boiling Point 120°C to 123°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3CH=CHCOCl
Refractive Index 1.4585 to 1.4615
Quantity 500 g
Beilstein 02, 411
Fieser 13,208
Specific Gravity 1.09
Solubility Information Solubility in water: decomposes
Formula Weight 104.54
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material trans-Crotonyl chloride, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-Crotonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.