accessibility menu, dialog, popup

UserName

CH3NHNH2S

CAS RN 625-53-6

IUPAC Name: ethylthiourea
Synonyms: N-ethylthiourea
Molecular Weight (g/mol): 104.17
Molecular Formula: C3H8N2S
InChi Key: GMEHFXXZSWDEDB-UHFFFAOYSA-N
SMILES: CCNC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results