missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylthiourea, 99%, Thermo Scientific Chemicals

Product Code. 11486476
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486476 250 g 250g
11466476 10 g 10g
11476476 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486476 Supplier Thermo Scientific Alfa Aesar Supplier No. B24761.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N-Ethylthiourea was used in the preparation of Re (III) complexes, which are precursors for the new rhenium complexes, potentially useful in nuclear medicine. It was also used in two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylthiourea was used in the preparation of Re (III) complexes, which are precursors for the new rhenium complexes, potentially useful in nuclear medicine. It was also used in two-step protocol for the first chemoselective cleavage of 2-hydroxy acid amides

Solubility
Soluble in water (24g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-53-6
Molecular Formula C3H8N2S
Molecular Weight (g/mol) 104.171
MDL Number MFCD00004939
InChI Key GMEHFXXZSWDEDB-UHFFFAOYSA-N
Synonym n-ethylthiourea, 1-ethylthiourea, 1-ethyl-2-thiourea, thiourea, ethyl, n-ethylthiocarbamide, ethyl thiourea, ethyl-2-thiourea, thiourea, n-ethyl, urea, 1-ethyl-2-thio, unii-u54or2e97e
PubChem CID 3032338
IUPAC Name ethylthiourea
SMILES CCNC(=S)N

Specifications

Melting Point 108°C to 112°C
Quantity 250 g
UN Number UN2811
Beilstein 1699551
Solubility Information Soluble in water (24g/L).
Formula Weight 104.17
Percent Purity 99%
Chemical Name or Material N-Ethylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.