accessibility menu, dialog, popup

UserName

H3CNSOOClOO

CAS RN 62522-63-8

IUPAC Name: 3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride
Synonyms: 3-methyl-2-oxo-1,3-benzoxazole-6-sulfonyl chloride
Molecular Weight (g/mol): 247.65
Molecular Formula: C8H6ClNO4S
InChi Key: MRSCMVKREGBNOW-UHFFFAOYSA-N
SMILES: CN1C(=O)OC2=CC(=CC=C12)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results