missing translation for 'onlineSavingsMsg'
Learn More

3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10386754
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10386754 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386754 Supplier Thermo Scientific Maybridge Supplier No. CC76703CB
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 62522-63-8
Molecular Formula C8H6ClNO4S
Molecular Weight (g/mol) 247.65
MDL Number MFCD03033702
InChI Key MRSCMVKREGBNOW-UHFFFAOYSA-N
Synonym 3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 3-methyl-2-oxo-2,3-dihydrobenzo d oxazole-6-sulfonyl chloride, 3-methyl-2-oxo-2,3-dihydro-benzooxazole-6-sulfonyl chloride, 6-benzoxazolesulfonyl chloride, 2,3-dihydro-3-methyl-2-oxo, 2,3-dihydro-3-methyl-2-oxo-6-benzoxazolesulfonyl chloride, 6-chlorosulfonyl-3-methylbenzoxazolin-2-one, 6-chlorosulfonyl-3-methyl-3-hydrobenzoxazol-2-one, pubchem5602, acmc-209n4g, 3-methyl-6-chlorosulfonylbenzoxazolone
PubChem CID 2772060
SMILES CN1C(=O)OC2=CC(=CC=C12)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride
Formula Weight 247.66
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.