accessibility menu, dialog, popup

UserName

CH3NNBrCH3

CAS RN 62546-27-4

IUPAC Name: 1-(4-bromophenyl)-3,5-dimethyl-1H-pyrazole
Synonyms: 1-(4-bromophenyl)-3,5-dimethylpyrazole
Molecular Weight (g/mol): 251.13
Molecular Formula: C11H11BrN2
InChi Key: ATCMNDCOEJOOSF-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results