accessibility menu, dialog, popup

UserName

H3CCH3OOCl

CAS RN 6258-30-6

IUPAC Name: 2-(4-chlorophenyl)-2-methylpropanoate
Synonyms: 2-(4-chlorophenyl)-2-methylpropanoate
Molecular Weight (g/mol): 197.64
Molecular Formula: C10H10ClO2
InChi Key: SSFDAZXGUKDEAH-UHFFFAOYSA-M
SMILES: CC(C)(C([O-])=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results