accessibility menu, dialog, popup

UserName

CAS RN 626-04-0

IUPAC Name: benzene-1,2-dithiol
Synonyms: 1,2-benzenedithiol
Molecular Weight (g/mol): 142.23
Molecular Formula: C6H6S2
InChi Key: JRNVQLOKVMWBFR-UHFFFAOYSA-N
SMILES: SC1=CC=CC=C1S
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)