accessibility menu, dialog, popup

UserName

CAS RN 626-58-4

IUPAC Name: 4-methylpiperidine
Synonyms: piperidine, 4-methyl-
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: UZOFELREXGAFOI-UHFFFAOYSA-N
SMILES: CC1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)