missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpiperidine, TRC

Product Code. 30467123
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467123 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467123 Supplier Toronto Research Chemicals Supplier No. TRCM32566510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylpiperidine
CAS 626-58-4
Molecular Formula C6H13N
Quantity 10 g
Synonym 4-Pipecoline, 4-Methylpiperidine, NSC 126910, NSC 66495, gamma-Pipecoline
InChI Formula InChI=1S/C6H13N/c1-6-2-4-7-5-3-6/h6-7H,2-5H2,1H3
SMILES CC1CCNCC1
IUPAC Name 4-methylpiperidine
Molecular Weight (g/mol) 99.17
Formula Weight 99.1048
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.