accessibility menu, dialog, popup

UserName

CAS RN 627-19-0

CH3HC

CAS RN 627-19-0

IUPAC Name: pent-1-yne
Synonyms: 1-pentyne
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: IBXNCJKFFQIKKY-UHFFFAOYSA-N
SMILES: CCCC#C
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)