missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyne, 99%

Product Code. 10686291 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686291 25 g Glass bottle 25g
10105403 5 g Glass bottle 5g
10638324 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686291 Supplier Thermo Scientific Acros Supplier No. 303820250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 627-19-0 | C5H8 | 68.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-19-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009469
InChI Key IBXNCJKFFQIKKY-UHFFFAOYSA-N
Synonym 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
PubChem CID 12309
IUPAC Name pent-1-yne
SMILES CCCC#C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -106.0°C to +105.0°C
Color Colorless to Yellow
Density 0.6900g/mL
Boiling Point 40.0°C
Flash Point <−34°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2CH2C≡CH
Refractive Index 1.3840 to 1.3880
Quantity 25 g
Specific Gravity 0.69
Formula Weight 68.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Pentyne, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.