accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 627-21-4

IUPAC Name: pent-2-yne
Synonyms: 2-pentyne
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: NKTDTMONXHODTI-UHFFFAOYSA-N
SMILES: CCC#CC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)