missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyne, 98%, ACROS Organics™

Product Code. 10319120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10319120 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10319120 Supplier Thermo Scientific Acros Supplier No. 307380050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 627-21-4
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.71g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H8
Linear Formula CH5CCCH3
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
MDL Number MFCD00009380
Quantity 5 g
Synonym 2-pentyne, ethylmethylacetylene, unii-57ng6hki9d, 1-ethyl-2-methylacetylene, 57ng6hki9d, acmc-209n7k, c2h5c.$.cch3, 2-pentyne 6ci,7ci,8ci,9ci
InChI Key NKTDTMONXHODTI-UHFFFAOYSA-N
SMILES CCC#CC
IUPAC Name pent-2-yne
Molecular Weight (g/mol) 68.12
PubChem CID 12310
Formula Weight 68.12
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Pentyne, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.