accessibility menu, dialog, popup

UserName

HOOClNOO

CAS RN 6280-88-2

IUPAC Name: 4-chloro-2-nitrobenzoic acid
Synonyms: 2-nitro-4-chlorobenzoic acid
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: JAHIPDTWWVYVRV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results