accessibility menu, dialog, popup

UserName

CAS RN 62899-78-9

ClClOHNNH2

CAS RN 62899-78-9

IUPAC Name: 3,5-dichlorobenzohydrazide
Synonyms: 3,5-dichlorobenzohydrazide
Molecular Weight (g/mol): 205.04
Molecular Formula: C7H6Cl2N2O
InChi Key: DNISXKBKTSHNKL-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results