accessibility menu, dialog, popup

UserName

CAS RN 629-19-6

IUPAC Name: 1-(propyldisulfanyl)propane
Synonyms: propyl disulfide
Molecular Weight (g/mol): 150.3
Molecular Formula: C6H14S2
InChi Key: ALVPFGSHPUPROW-UHFFFAOYSA-N
SMILES: CCCSSCCC
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)