missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl disulfide, 99%

Product Code. 11362327
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362327 100 g 100g
11352327 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09881.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-19-6 | C6H14S2 | 150.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-19-6
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00009378
InChI Key ALVPFGSHPUPROW-UHFFFAOYSA-N
Synonym dipropyl disulfide, propyl disulfide, disulfide, dipropyl, n-propyl disulfide, di-n-propyl disulfide, 4,5-dithiaoctane, dipropyl disulphide, 1-propyldisulfanyl propane, propyldithiopropane, 1,1'-dithiodipropane
PubChem CID 12377
ChEBI CHEBI:45758
IUPAC Name 1-(propyldisulfanyl)propane
SMILES CCCSSCCC

Specifications

Melting Point -86°C
Density 0.96
Boiling Point 193°C to 194°C
Flash Point 64°C (147°F)
Odor Unpleasant
Refractive Index 1.4967
Quantity 100 g
UN Number UN3082
Beilstein 969200
Formula Weight 150.31
Percent Purity 99%
Chemical Name or Material Di-n-propyl disulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.