accessibility menu, dialog, popup

UserName

CAS RN 629-27-6

H3CI

CAS RN 629-27-6

IUPAC Name: 1-iodooctane
Synonyms: 1-iodooctane
Molecular Weight (g/mol): 240.13
Molecular Formula: C8H17I
InChi Key: UWLHSHAHTBJTBA-UHFFFAOYSA-N
SMILES: CCCCCCCCI
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)