missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctane, 98%, stabilized

Product Code. 11974501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11974501 500 g Glass bottle 500g
10361251 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11974501 Supplier Thermo Scientific Acros Supplier No. 412465000
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-27-6 | C8H17I | 240.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.13
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -46°C to 45°C
Color Colorless to Amber
Density 1.3300g/mL
Boiling Point 225.0°C to 226.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7I
Refractive Index 1.4878
Quantity 500 g
Beilstein 01,16
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Stabilizer copper
Formula Weight 240.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iodooctane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodooctane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.