accessibility menu, dialog, popup

UserName

CAS RN 629-96-9

H3COH

CAS RN 629-96-9

IUPAC Name: icosan-1-ol
Synonyms: arachidyl alcohol
Molecular Weight (g/mol): 298.56
Molecular Formula: C20H42O
InChi Key: BTFJIXJJCSYFAL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)