missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Eicosanol, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Arachidyl Alcohol |
| CAS | 629-96-9 |
| Molecular Formula | C20 H42 O |
| Quantity | 250 mg |
| Synonym | 1-Eicosanol, Eicosanol (7CI), Arachic alcohol, Arachidic alcohol, Arachidyl alcohol, Eicosyl alcohol, Hainol 20SS, NSC 120887, Nacol C 20, n-1-Eicosanol, n-Eicosanol |
| InChI Formula | InChI=1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3 |
| SMILES | CCCCCCCCCCCCCCCCCCCCO |
| IUPAC Name | icosan-1-ol |
| Molecular Weight (g/mol) | 298.55 |
| Formula Weight | 298.3236 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction