accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 6304-16-1

IUPAC Name: 1-(pyridin-4-yl)propan-2-one
Synonyms: 1-(pyridin-4-yl)propan-2-one
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: ILRVKOYYFFNXDB-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results