missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (4-Pyridyl)acetone, 98%

Product Code. 11399445
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399445 5 g 5g
11389445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399445 Supplier Alfa Aesar™ Supplier No. L18667.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
(4-Pyridyl)acetone is a reactant in the synthesis of Milrinone (M344680), a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity. Cardiotonic.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6304-16-1
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00129043
InChI Key ILRVKOYYFFNXDB-UHFFFAOYSA-N
Synonym 4-pyridyl acetone, 1-4-pyridinyl-2-propanone, 1-pyridin-4-yl propan-2-one, 1-4-pyridyl acetone, 4-acetonylpyridine, 4-pyridinylacetone, 4-pyridylacetone, unii-r6onm9f2au, 2-propanone, 1-4-pyridinyl
PubChem CID 80552
IUPAC Name 1-pyridin-4-ylpropan-2-one
SMILES CC(=O)CC1=CC=NC=C1

Specifications

Melting Point 13°C
Boiling Point 143°C (20 mmHg)
Refractive Index 1.5225
Quantity 5 g
Beilstein 110960
Solubility Information Slightly soluble in water.
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material (4-Pyridyl)acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.