accessibility menu, dialog, popup

UserName

CAS RN 6304-33-2

N

CAS RN 6304-33-2

IUPAC Name: triphenylprop-2-enenitrile
Synonyms: triphenylacrylonitrile 2,3,3-trifenylakrylonitril
Molecular Weight (g/mol): 281.36
Molecular Formula: C21H15N
InChi Key: OPACMJHTQWVSKW-UHFFFAOYSA-N
SMILES: N#CC(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results