accessibility menu, dialog, popup

UserName

CAS RN 631-57-2

IUPAC Name: acetyl cyanide
Synonyms: acetyl cyanide
Molecular Weight (g/mol): 69.06
Molecular Formula: C3H3NO
InChi Key: QLDHWVVRQCGZLE-UHFFFAOYSA-N
SMILES: CC(=O)C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results