missing translation for 'onlineSavingsMsg'
Learn More

Pyruvonitrile, tech. 90%

Product Code. 15432799
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432799 5 g 5g
15422799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432799 Supplier Thermo Scientific Alfa Aesar Supplier No. H66636.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 631-57-2 | C3H3NO | 69.063 g/mol

Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

Solubility
Soluble in ether and acetonitrile.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-57-2
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00001838
InChI Key QLDHWVVRQCGZLE-UHFFFAOYSA-N
Synonym pyruvonitrile, propanenitrile, 2-oxo, oxopropionitrile, 2-oxopropiononitrile, 2-oxopropionitrile, oxypropionitrile, pyruvic acid nitrile, unii-ulo441649v, accn, .alpha.-oxopropionitrile
PubChem CID 69430
IUPAC Name acetyl cyanide
SMILES CC(=O)C#N

Specifications

Color Colorless to Brown
Density 0.974 g/mL
Boiling Point 92°C to 93°C
Flash Point 14°C (57°F)
Linear Formula CH3COCN
Refractive Index 1.3755
Quantity 5 g
UN Number UN3273
Beilstein 1737633
Solubility Information Soluble in ether and acetonitrile.
Formula Weight 69.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Pyruvonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.