CAS RN 6317-18-6
IUPAC Name:
{[(cyanosulfanyl)methyl]sulfanyl}carbonitrile
Synonyms:
methylene bisthiocyanate
Molecular Weight (g/mol):
130.18
Molecular Formula:
C3H2N2S2
InChi Key:
JWZXKXIUSSIAMR-UHFFFAOYSA-N
SMILES:
N#CSCSC#N
1
–
2
of
2
results
1
Methylene Dithiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methylene Dithiocyanate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.