missing translation for 'onlineSavingsMsg'
Learn More

Methylene Dithiocyanate, TRC

Product Code. 30475166
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475166 Supplier Toronto Research Chemicals Supplier No. TRCM3041155G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylene dithiocyanate
CAS 6317-18-6
Molecular Formula C3 H2 N2 S2
Quantity 5 g
Synonym Methylene Dithiocyanate, Thiocyanic acid','C,C'-methylene ester (ACI), Thiocyanic acid','methylene ester (6CI','7CI','8CI','9CI), Antiblu 3737, Antiblu 3738, Basiment 540, Bis(thiocyanato)methane, Busan 110, Dithiocyanomethane, Kilstain, MBT, Metasol T 10, Methylene bis(thiocyanate), Methylene dithiocyanate, Methylene thiocyanate, Methylenedirhodanide, Nalco 5793, Nalco D 2303, Nalfloc N 206, NSC 40464, Proxel MB, Slimicide MC, V 709, WT 307B
InChI Formula InChI=1S/C3H2N2S2/c4-1-6-3-7-2-5/h3H2
SMILES N#CSCSC#N
IUPAC Name thiocyanatomethyl thiocyanate
Molecular Weight (g/mol) 130.19
Formula Weight 129.9659
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.