accessibility menu, dialog, popup

UserName

NH2OHHOHCl

CAS RN 6318-56-5

IUPAC Name: 5-aminobenzene-1,3-diol hydrochloridyl
Synonyms: 5-aminobenzene-1,3-diol hydrochloridyl
Molecular Weight (g/mol): 160.58
Molecular Formula: C6H7ClNO2
InChi Key: BEPMWVUSEBWNCW-UHFFFAOYSA-N
SMILES: [Cl].NC1=CC(O)=CC(O)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results