accessibility menu, dialog, popup

UserName

CAS RN 6320-02-1

IUPAC Name: 2-bromobenzene-1-thiol
Synonyms: 2-bromobenzenethiol
Molecular Weight (g/mol): 189.07
Molecular Formula: C6H5BrS
InChi Key: YUQUNWNSQDULTI-UHFFFAOYSA-N
SMILES: SC1=CC=CC=C1Br
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)