accessibility menu, dialog, popup

UserName

ClClICl

CAS RN 6324-50-1

IUPAC Name: 1,3,5-trichloro-2-iodobenzene
Synonyms: 1,3,5-trichloro-2-iodobenzene
Molecular Weight (g/mol): 307.34
Molecular Formula: C6H2Cl3I
InChi Key: XXFFSBRDPQFIPO-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(I)C(Cl)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results