CAS RN 6325-91-3
CAS RN 6325-91-3
IUPAC Name:
5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms:
5-nitro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol):
195.2
Molecular Formula:
C7H5N3O2S
InChi Key:
YPXQSGWOGQPLQO-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C2NC(=S)NC2=C1
1
–
1
of
1
results
1
2-Mercapto-5-nitrobenzimidazole, 97%
CAS: 6325-91-3 Molecular Formula: C7H5N3O2S Molecular Weight (g/mol): 195.20 MDL Number: MFCD00804727 InChI Key: YPXQSGWOGQPLQO-UHFFFAOYSA-N Synonym: 2-mercapto-5-nitrobenzimidazole,5-nitro-2-benzimidazolethiol,5-nitro-2-mercaptobenzimidazole,5-nitro-1h-benzo d imidazole-2-thiol,2-benzimidazolethiol, 5-nitro,2-benzimidazolinethione, 5-nitro,2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro,6-nitro-1h-benzimidazole-2-thiol,5-nitro-1h-benzimidazole-2-thiol,5-nitro-3-hydrobenzimidazole-2-thione PubChem CID: 3717598 IUPAC Name: 5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione SMILES: [O-][N+](=O)C1=CC=C2NC(=S)NC2=C1