missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-nitrobenzimidazole, 97%

Product Code. 11466086
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11466086

missing translation for 'mfr': Thermo Scientific Alfa Aesar B22358.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6325-91-3 | C7H5N3O2S | 195.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6325-91-3
Molecular Formula C7H5N3O2S
Molecular Weight (g/mol) 195.20
MDL Number MFCD00804727
InChI Key YPXQSGWOGQPLQO-UHFFFAOYSA-N
Synonym 2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione
PubChem CID 3717598
IUPAC Name 5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
SMILES [O-][N+](=O)C1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point ∼273°C (decomposition)
Quantity 100 g
Beilstein 172127
Formula Weight 195.2
Percent Purity 97%
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.