CAS RN 6337-56-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy-2,6-dimethylacetanilide, TRC
CAS: 6337-56-0 Molecular Formula: C10H13NO2 Molecular Weight (g/mol): 179.22 Synonym: N-(4-Hydroxy-2,6-dimethylphenyl)acetamide,4'-Hydroxy-2',6'-acetoxylidide,2,6-Dimethylparacetamol,2,6-Dimethylacetaminophen,NSC 38035; IUPAC Name: N-(4-hydroxy-2,6-dimethylphenyl)acetamide SMILES: CC(=O)Nc1c(C)cc(O)cc1C