accessibility menu, dialog, popup

UserName

H3CONH2O

CAS RN 6343-93-7

IUPAC Name: 2-(4-methoxyphenyl)acetamide
Synonyms: 2-(4-methoxyphenyl)acetamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: OLKQIWCQICCYQS-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(N)=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results