accessibility menu, dialog, popup

UserName

CAS RN 635-51-8

IUPAC Name: 2-phenylbutanedioic acid
Synonyms: 2-phenylbutanedioic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: LVFFZQQWIZURIO-UHFFFAOYNA-N
SMILES: OC(=O)CC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)